Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piperonylamine, 97%, Thermo Scientific™

Catalog No. p-185501
Click to view available options
Quantity:
25 mL
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2620-50-0
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00005840
InChI Key ZILSBZLQGRBMOR-UHFFFAOYSA-N
Synonym piperonylamine, benzo d 1,3 dioxol-5-ylmethanamine, 3,4-methylenedioxybenzylamine, 1,3-benzodioxol-5-ylmethylamine, 1,3-benzodioxole-5-methanamine, 2h-1,3-benzodioxol-5-ylmethanamine, 3,4-methylenedioxy benzylamine, benzo-1,3-dioxole-5-methylamine, 1-1,3-benzodioxol-5-yl methanamine, 5-aminomethyl-1,3-benzodioxole
PubChem CID 75799
IUPAC Name 1,3-benzodioxol-5-ylmethanamine
SMILES NCC1=CC=C2OCOC2=C1

Specifications

Color Yellow
Density 1.2100g/mL
Boiling Point 138.0°C to 139.0°C (13.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Refractive Index 1.5625 to 1.5645
Quantity 25 mL
Beilstein 19,I,764
Specific Gravity 1.21
Solubility Information Solubility in water: 0.9%. Other solubilities: soluble in alcohol, acetone and toluene
Formula Weight 151.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Piperonylamine, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 220-056-1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.