Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIPES, Sesquisodium Sodium Salt, Fisher BioReagents

Catalog No. p-27672 Shop All Fisherbrand Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Poly Bottle

Commonly used buffering agent

This biological buffer has a usable pH range of 6.1 to 7.5.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100037-69-2
Molecular Formula C16H33N4Na3O12S4
Molecular Weight (g/mol) 670.665
InChI Key FPDACHMSOMILQI-UHFFFAOYSA-K
Synonym 1, 4-Piperazinediethanesulfonic acid sesquisodium salt, Piperazine-1, 4-bis(2-ethanesulfonic acid) sesquisodium salt
PubChem CID 3086462
IUPAC Name trisodium;2-[4-(2-sulfoethyl)piperazin-1-yl]ethanesulfonate;2-[4-(2-sulfonatoethyl)piperazin-1-yl]ethanesulfonate
SMILES C1CN(CCN1CCS(=O)(=O)O)CCS(=O)(=O)[O-].C1CN(CCN1CCS(=O)(=O)[O-])CCS(=O)(=O)[O-].[Na+].[Na+].[Na+]

Specifications

Melting Point 300°C
Color White
pH 6.5 to 7.1
Physical Form Powder or Solid
Quantity 100 g
DNase DNase free
Assay Percent Range ≥95 %
Identification Pass Test
Packaging Poly Bottle
Protease Protease free
Formula Weight 670.68
Percent Purity ≥95%
Grade DNase- and RNase-Free
Chemical Name or Material PIPES, Sesquisodium Salt
Show More Show Less
Recommended Storage RT
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.