Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Heathrow Scientific Biohazard Pipet Keeper XL Container, CS 50
Shop All Fisherbrand Products
Click to view available options
Dimensions (L x W x H):
8.89 x 11.43 x 41.91 cm (3.5 x 4.5 x 16.5 in.)
Description
- The Pipet Keeper XL provides a larger, secure solution for collecting lab contaminants while keeping workstations clean and compliant
- Holds biohazardous waste up to 16 in. length
- Sturdy paperboard construction with visible biohazard symbols
- Self-locking top and re-closable flaps for secure containment
- Easily disposable after autoclaving per regulations
- Cost-effective and user-friendly design
Specifications
Specifications
| Capacity (English) | 4200 cc |
| Color | White, Orange, Blue |
| Dimensions (L x W x H) | 8.89 x 11.43 x 41.91 cm (3.5 x 4.5 x 16.5 in.) |
| Quantity | 50/Cs. |
| Autoclavable | Autoclavable |
| Includes | Pipet Keeper |
| Material | Plastic-lined Paperboard |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction