Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Pipet Tips
SCI_ED

Catalog No. S66805A Shop All Fisherbrand Products
Change view
Click to view available options
Volume (Metric):
1 to 200 μL
Tip Style:
Standard
Catalog No. Volume (Metric) Tip Style
S66805A 1 to 200 μL Standard
1 options

Catalog No. S66805A

General purpose for universal applications

  • Reference tips with graduation marks measure variable amounts with fixed volume pipetter
  • Gel loading tips with fine points for loading electrophoresis gels

Order Info

Packaged in bulk packs of 1000, except the sterile gel loading tips (S63216) which are packaged in five racks of 204.

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 1 to 200 μL
Sterility Non-sterile
Color Yellow
Autoclavable Autoclavable
Length (English) 2 in.
Length (Metric) 5cm
Clean Claims DNase, RNase, DNA, and Pyrogen Free
Tip Style Standard
Product Type `
For Use With (Equipment) Universal Fit
Graduated Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.