Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ PIPETMAN™ Classic Pipette Trade-In PROMO

Catalog No. p-7182023
Click to view available options
Volume (Metric):
0.2 to 2 μL
1 to 10 μL
1 to 10 mL
1 to 5 mL
2 to 20 μL
20 to 100 μL
20 to 200 μL
200 to 1000 μL

Send in your used or broken pipettes and save on select Gilson models. All used brands and models qualify and a pre-postage bag will be included with the order. Refurbished pipettes will be donated to Seeding Labs, who distributes to labs and universities around the world who need it most.

  • Fully adjustable, air-displacement pipette with selected volume range
  • Volume shown on digital indicator (volumeter)
  • Autoclavable tip holder and tip ejector help to avoid cross-contamination
  • Each pipette bears engraved identification number for traceability

Specifications

Accuracy ±0.024, ±12 μL
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color White/Blue
Dispensing Speed Manual
Disposable Reusable
For Use With D-10, DL10, DF10ST, DFL10ST Tip
Grip Type Ergonomic
Material PVDF, Stainless Steel
Model P2
Number of Channels 1
Product Line Pipetman Classic
Programmability No
Sterility Non-sterile
Type Pipette
Volume (Metric) 0.2 to 2 μL
For Use With (Application) PCR, DNA sequencing
For Use With (Equipment) D10, DL10, DF10ST, DFL10ST tips
Color Code Red
Fit for Purpose Certified Quality
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.