Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ PIPETMAN™ L Mechanical Pipets PROMO

Catalog No. p-7160289
Click to view available options
Volume (Metric):
0.2 to 2 μL
1 to 10 μL
10 to 100 μL
100 to 1000 μL
2 to 20 μL
20 to 200 μL

Gilson™ PIPETMAN™ L Mechanical Pipets are a lightweight pipet with lockable volume and low forces.

  • Redesigned handle fits naturally in user's hand, no matter the size
  • Volume locking system guarantees no accidental change in volume while pipetting
  • Adjustable tip ejector provides comfort for right- and left-handed users
  • 2D identification code allows easy tracking of product information
  • New piston assembly designed to drastically improve pipetting and purge forces
  • Redesigned tip ejector shape and internal mechanism offers reduced tip ejection forces by 50%

Specifications

Accuracy ±0.024, ±0.025, ±0.027, ±0.03 μL
Compatible Tips D10, DL10, DF10ST, DFL10ST tip
Color Orange
Model P2L
Product Line Pipetman L
Type Mechanical Pipette
Volume (Metric) 0.2 to 2 μL
Finger Style Ergonomic
For Use With (Application) Transporting a measured volume of liquid
Includes Stainless Steel tip ejector
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.