Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gilson™ PIPETMANTM L Multichannel Pipettes
GREENER_CHOICE

Catalog No. p-9217001 Shop All Gilson Products
Click to view available options
Number of Channels:
8
12

Enhanced from the legendary PIPETMANTM Classic with a newly designed body for improved comfort and a state-of-the-art volume-locking mechanism for precision.

Optimized Comfort

  • One of the most comfortable models in the Gilson portfolio
  • Ideal weight and handle shape
  • Natural fit of the pipette allows for a relaxed grip to make pipetting less fatiguing
  • Features an updated piston assembly with more responsive springs to reduce pipetting forces and decrease the risk of repetitive strain injuries
  • Adjustable tip ejector for right- or left-handedness, whichever is most comfortable

Optimized Safety and Precision

  • Volume-locking ensures no accidental volume change during pipetting cycles, which could alter the accuracy and precision of results
  • Patented locking system fully disconnects the operating rod from the counter assembly during pipetting
  • Once locked into place, the volume cannot be accidentally changed to prevent accidental volume drift during pipetting tasks
  • Variable volume models are fully autoclavable to minimize cross-contamination
  • No disassembly required for maximum convenience

Optimized Traceability

  • Features a 2D identification bar code for easy traceability
  • Particularly suited for enhanced GLP compliance
  • Color-coded push button corresponds in color to Gilson PIPETMANTM Tip packages
  • Using a universal scanner download the product part number, date of manufacturing, and serial number

Specifications

Compatible Tips PIPETMAN DIAMOND Tips D1200, DF1200ST
Adjustable Tip Spacing No
Autoclavable Autoclavable
Color Dark Blue
Dispensing Speed Manual
Disposable Reusable
For Use With Pipette Tip 500 to 5000 μL
Grip Type Ergonomic
Material Polypropylene, PVDF, PBT
Model Pipetman™ L
Number of Channels 8
Product Line Pipetman
Programmability No
Sterility Non-sterile
Type Manual Air Displacement Pipette
Volume (Metric) 100 to 1200 μL
Quantity 1
System Type Metal Ejector
Width (English) 5.98 in
Color Code Blue
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.