Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Gilson™ PIPETMANTM L Multichannel Pipette Trade-In PROMO
Shop All Gilson Products

Click to view available options
Color:
Green
Red
White
Yellow
Volume (Metric):
0.5 to 10 μL
2 to 20 μL
20 to 200 μL
20 to 300 μL
Number of Channels:
8
12
Description
- Gilson-patented volume locking system guarantees no accidental change in volume while pipetting
- Adjustable tip ejector provides comfort for right- and left-handed users
- Redesigned handle fits naturally in your hand, regardless of the size
- Unique internal design applies even, equal pressure from either side of the pipettes, rather than on the center of the channels like many other multichannel pipettes do
- 2D identification code tracks product reference number, date of manufacture, serial number, and maximum volume of pipette
- Available in 8- or 12- channels, in volumes ranging from 0.50 to 300 μL
Specifications
Specifications
| Compatible Tips | PIPETMAN DIAMOND Tips D200, D300, DF200ST, DF300ST |
| Adjustable Tip Spacing | No |
| Autoclavable | Autoclavable |
| Color | Green |
| Dispensing Speed | Manual |
| Disposable | Reusable |
| For Use With | Pipette Tip 50 to 1000 μL |
| Grip Type | Ergonomic |
| Material | Polypropylene |
| Model | P8x300L |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction