Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Poltex Pipette Tip Receptacle
SDP

Catalog No. 22000758
Click to view available options
Includes:
Magnet Mount
VHB Tape
Wall Mount
This item is not returnable. View return policy

Catalog No. 22-000-758

Supplier: Poltex TIPDISPM

Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Receptacle for used pipette tips.

  • Back plate mounts to wall, inside of hood, etc.
  • Front receptacle lifts off for easy emptying/cleaning.
  • Choose from VHB tape mount (very high bond tape included), or magnet mount (powerful magnets included).

Specifications

Product Type Pipet Rack
Includes Magnet Mount
Array 1 x 1
Autoclavable Not Autoclavable
Color Clear
Holds Pipettes
Material PETG
Positions 1
Shape Rectangular, Rounded Corners
Stackable No
Sterility Non-sterile
For Use With (Equipment) Pipettes
Length (Metric) 11 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.