Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIPOX Monoclonal Antibody (3D1)
GREENER_CHOICE

Catalog No. PIMA521892
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA521892 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA521892 Supplier Invitrogen™ Supplier No. MA521892
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: IGDVQILSSF VRDHLPDLKP EPAVIESCMY TNTPDEQFIL DRHPKYDNIV IGAGFSGHGF KLAPVVGKIL YELSMKLTPS YDLAPFRISR FPG.

Metabolizes sarcosine, L-pipecolic acid and L-proline.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIPOX
Applications ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 3D1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene Pipox
Gene Accession No. Q9P0Z9
Gene Alias L-pipecolate oxidase; L-pipecolic acid oxidase; LPIPOX; Peroxisomal sarcosine oxidase; pipecolic acid and sarcosine oxidase; pipecolic acid oxidase; PIPOX; Pso
Gene Symbols Pipox
Host Species Mouse
Immunogen PIPOX (AAH27622.1, 292 a.a. ~ 384 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51268
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.