Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIPOX Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550084
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550084 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550084 Supplier Invitrogen™ Supplier No. PA550084
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total PIPOX protein.

Metabolizes sarcosine, L-pipecolic acid and L-proline.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIPOX
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Pipox
Gene Accession No. Q9D826, Q9P0Z9
Gene Alias L-pipecolate oxidase; L-pipecolic acid oxidase; LPIPOX; Peroxisomal sarcosine oxidase; pipecolic acid and sarcosine oxidase; pipecolic acid oxidase; PIPOX; Pso
Gene Symbols Pipox
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human PIPOX.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19193, 51268
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.