Learn More
Invitrogen™ PIR-A/PIR-B Monoclonal Antibody (404127)

Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
Specifications
| Antigen | PIR-A/PIR-B |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 404127 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 5% trehalose and No Preservative |
| Gene | Pira1 |
| Gene Accession No. | P97484 |
| Gene Alias | 6M21; CD85k; cell-surface glycoprotein p91; Gp49b; Gp91; leukocyte immunoglobulin-like receptor 3; leukocyte immunoglobulin-like receptor subfamily B member 3; leukocyte immunoglobulin-like receptor, subfamily B (with TM and ITIM domains), member 3; Lilrb3; Lilrb4; LIR-3; Ly89; paired Ig-like receptor B; paired immunoglobulin-like receptor; paired immunoglobulin-like receptor B; paired-Ig-like receptor A1; Pir; Pira1; PIR-A1; Pirb; PIR-B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.