Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIR-B Antibody (326414), DyLight 594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB2754M
Description
PIR-B Monoclonal antibody specifically detects PIR-B in Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOFSpecifications
PIR-B | |
Monoclonal | |
DyLight 594 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
11025 | |
Store at 4°C in the dark. | |
IgG2a |
Western Blot, Flow Cytometry, CyTOF | |
326414 | |
Western Blot, Flow Cytometry, CyTOF-ready | |
CD85a, HL9, ILT5, ILT-5, leukocyte immunoglobulin like receptor B3, LILRA6, LILRB3, LIR3, LIR-3, PIRB, PIR-B | |
Mouse myeloma cell line NS0-derived recombinant mouse PIR-B, Gly24-Gly635, Accession # AAH26937 | |
0.1 mL | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction