Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pirh2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
$343.50 - $573.00
Specifications
Antigen | Pirh2 |
---|---|
Dilution | Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL |
Applications | Immunofluorescence |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Pirh2 Polyclonal antibody specifically detects Pirh2 in Human samples. It is validated for ImmunofluorescenceSpecifications
Pirh2 | |
Immunofluorescence | |
Unconjugated | |
Rabbit | |
Apoptosis, Breast Cancer, Cancer, p53 Pathway, Tumor Suppressors, Ubiquitin Proteasome Pathway, Zinc Finger | |
PBS, pH 7.2, 40% glycerol | |
25898 | |
IgG | |
Affinity purified |
Immunocytochemistry/Immunofluorescence 0.25 to 2 μg/mL | |
Polyclonal | |
Purified | |
RUO | |
Human | |
Androgen receptor N-terminal-interacting protein, androgen-receptor N-terminal-interacting protein, ARNIPRING finger protein 199, CHIMPRNF199DKFZp586C1620, CH-rich interacting match with PLAG1, CH-rich-interacting match with PLAG1, E3 ubiquitin-protein ligase Pirh2, EC 6.3.2.-, hARNIP, hPirh2, PIRH2, PRO1996, ring finger and CHY zinc finger domain containing 1, RING finger and CHY zinc finger domain-containing protein 1, Zinc finger protein 363p53-induced RING-H2 protein, ZNF363 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: TAREDGASGQERGQRGCEHYDRGCLLKAPCCDKLYTCRLCHDNNEDHQLDRFKVKEVQCINCEKIQHAQQTCEECSTLFGEYY | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title