Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pirh2 Recombinant Rabbit Monoclonal Antibody (4Q5H9)
GREENER_CHOICE

Catalog No. PIMA537910
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537910 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537910 Supplier Invitrogen™ Supplier No. MA537910
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive Samples: HCT116 Immunogen sequence: HVLPCGHLLH RTCYEEMLKE GYRCPLCMHS ALDMTRYWRQ LDDEVAQTPM PSEYQNMTVD ILCNDCNGRS TVQFHILGMK CKICESYNTA QAGGRRISLD QQ.

The protein encoded by this gene has ubiquitin-protein ligase activity. This protein binds with p53 and promotes the ubiquitin-mediated proteosomal degradation of p53. This gene is oncogenic because loss of p53 function contributes directly to malignant tumor development. Transcription of this gene is regulated by p53. Alternative splicing results in multiple transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pirh2
Applications ELISA, Western Blot
Classification Recombinant Monoclonal
Clone 4Q5H9
Concentration 1.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene RCHY1
Gene Accession No. Q96PM5, Q9CR50
Gene Alias Androgen receptor N-terminal-interacting protein; androgen-receptor N-terminal-interacting protein; ARNIP; Chimp; CH-rich interacting match with PLAG1; CH-rich-interacting match with PLAG1; DKFZp586C1620; E3 ubiquitin-protein ligase Pirh2; h PRO1996; hPirh2; p53-induced protein with a RING-H2 domain; p53-induced RING-H2 protein; PIRH2; PIRH2E; PIRH2F; PRO1996; RCHY1; ring finger and CHY zinc finger domain containing 1; ring finger and CHY zinc finger domain containing 1, E3 ubiquitin protein ligase; RING finger and CHY zinc finger domain-containing protein 1; RING finger protein 199; RING-type E3 ubiquitin transferase RCHY1; RNF199; ZCHY; Zfp363; Zinc finger protein 363; zinc finger, CHY-type; Znf363
Gene Symbols RCHY1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 160-261 of human RCHY1 (Q96PM5).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25898, 68098
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.