Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIRIN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551441
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551441 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551441 Supplier Invitrogen™ Supplier No. PA551441
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: TVSYLLEGGS MAHEDFCGHT GKMNPGDLQW MTAGRGILHA EMPCSEEPAH GLQLWVNLRS SEKMVEPQYQ ELKSEEIPKP SKDGVTVAVI SGEALGIKSK VYTRTPTLYL DF Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 97%.

This gene encodes a member of the cupin superfamily. The encoded protein is an Fe(II)-containing nuclear protein expressed in all tissues of the body and concentrated within dot-like subnuclear structures. Interactions with nuclear factor I/CCAAT box transcription factor as well as B cell lymphoma 3-encoded oncoprotein suggest the encoded protein may act as a transcriptional cofactor and be involved in the regulation of DNA transcription and replication. Alternatively spliced transcript variants have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIRIN
Applications Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PIR
Gene Accession No. O00625, Q5M827
Gene Alias 2310042L19Rik; iron-binding nuclear protein; PIR; Pirin; pirin (iron-binding nuclear protein); pirin like; pirinin; Pirnl; Probable quercetin 2,3-dioxygenase PIR; probable quercetinase
Gene Symbols PIR
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PIRIN. Recombinant protein control fragment (Product #RP-91913).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 363465, 8544
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.