Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIRT Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144340
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144340 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144340 Supplier Invitrogen™ Supplier No. PA5144340
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PIRT Polyclonal Antibody for ICC/IF, IHC (P), ELISA

PIRT is a key component of the TRPV1 complex that positively regulates TRPV1 activity. It binds various phosphoinositide, including phosphatidylinositol 4, 5-bisphosphate (PIP2), but not phosphatidylinositol (PI). PIRT is a membrane protein with MW 15-25 kDa.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIRT
Applications ELISA, Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene PIRT
Gene Accession No. P0C851
Gene Alias A530088H08Rik; phosphoinositide interacting protein; phosphoinositide interacting regulator of transient receptor potential channels; phosphoinositide-interacting protein; phosphoinositide-interacting regulator of transient receptor potential channels; phosphoinositide-interacting regulator of TRPV1; Pirt; RGD1565284
Gene Symbols PIRT
Host Species Rabbit
Immunogen Recombinant Human Phosphoinositide-interacting protein (1-55AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644139
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.