Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PIRT Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981526
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1526 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1526 Supplier Bioss Supplier No. BS11623R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PIRT is a key component of the TRPV1 complex that positively regulates TRPV1 activity. It binds various phosphoinositide, including phosphatidylinositol 4, 5-bisphosphate (PIP2), but not phosphatidylinositol (PI). PIRT is a membrane protein with MW 15-25 kDa.

Specifications

Antigen PIRT
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PIRT
Gene Accession No. P0C851
Gene Alias phosphoinositide interacting regulator of transient receptor potential channels; Phosphoinositide-interacting protein; phosphoinositide-interacting regulator of transient receptor potential channels; phosphoinositide-interacting regulator of TRPV1; PIRT
Gene Symbols PIRT
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PIRT.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 644139
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.