Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PISD Monoclonal Antibody (OTI3B11)
GREENER_CHOICE

Catalog No. PIMA526860
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526860 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526860 Supplier Invitrogen™ Supplier No. MA526860
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

PISD Monoclonal Antibody for Western Blot, IHC (P)

Phosphatidylserine decarboxylases catalyze the formation of phosphatidylethanolamine (PE) by decarboxylation of phosphatidylserine (PS). Type I PSDs, such as PISD, are targeted to the inner mitochondrial membrane by an N-terminal targeting sequence. PISD also contains a conserved LGST motif that functions as an autocatalytic cleavage site where the proenzyme is split into mature alpha and beta subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PISD
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3B11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PISD
Gene Accession No. Q9UG56
Gene Alias 9030221M09Rik; DJ858B16; dJ858B16.2; DKFZp566G2246; phosphatidylserine decarboxylase; Phosphatidylserine decarboxylase alpha chain; Phosphatidylserine decarboxylase beta chain; phosphatidylserine decarboxylase proenzyme; phosphatidylserine decarboxylase proenzyme, mitochondrial; phosphatidylserine decarboxylase proenzyme; phosphatidylserine decarboxylase proenzyme, mitochondrial; Pisd; PSD; PSDC; PSSC
Gene Symbols PISD
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 80-340 of PISD produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23761
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.