Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PISD Antibody (OTI4G5), DyLight 680, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273420FR
Description
PISD Monoclonal specifically detects PISD in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PISD | |
Monoclonal | |
DyLight 680 | |
PBS with 0.05% Sodium Azide | |
PISD | |
Recombinant protein fragment corresponding to amino acids 80-340 of human PISD (NP_055153) produced in E.coli. | |
0.1 mL | |
23761.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4G5 | |
Western Blot, Immunohistochemistry | |
DJ858B16, dJ858B16.2, DKFZp566G2246, EC 4.1.1.65, phosphatidylserine decarboxylase, phosphatidylserine decarboxylase proenzyme, PSD, PSDC, PSSC | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction