Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PISD Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556634
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556634 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556634 Supplier Invitrogen™ Supplier No. PA556634
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YKSVPTRLLS RAWGRLNQVE LPHWLRRPVY SLYIWTFGVN MKEAAVEDLH HYRNLSEFFR RKLKPQARPV CGLHSISPSD Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.

Phosphatidylserine decarboxylases catalyze the formation of phosphatidylethanolamine (PE) by decarboxylation of phosphatidylserine (PS). Type I PSDs, such as PISD, are targeted to the inner mitochondrial membrane by an N-terminal targeting sequence. PISD also contains a conserved LGST motif that functions as an autocatalytic cleavage site where the proenzyme is split into mature alpha and beta subunits.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PISD
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PISD
Gene Accession No. Q9UG56
Gene Alias 9030221M09Rik; DJ858B16; dJ858B16.2; DKFZp566G2246; phosphatidylserine decarboxylase; Phosphatidylserine decarboxylase alpha chain; Phosphatidylserine decarboxylase beta chain; phosphatidylserine decarboxylase proenzyme; phosphatidylserine decarboxylase proenzyme, mitochondrial; phosphatidylserine decarboxylase proenzyme; phosphatidylserine decarboxylase proenzyme, mitochondrial; Pisd; PSD; PSDC; PSSC
Gene Symbols PISD
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PISD. Recombinant protein control fragment (Product #RP-95350).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23761
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.