Learn More
StatLab PiSmart S1 Single Hopper Thermal Transfer Slide Printer
Click to view available options
Product Type:
PiSmart S1 Printer
PiSmart S1 Printer Kit
Description
- Designed for on-demand printing, the PiSmart S1 Single Hopper Thermal Transfer
- Slide Printer can deliver the first printed slide in as little as 3 seconds
- Superior print quality produces dark, durable print with excellent contrast and supports up to 200 characters: 300 dpi.
- Users have the ability to easily swap hoppers, as well as bypass the hopper and manually drop in a different type of slide
- Integrated label designer no additional software needed
- Small footprint is ideal for on-demand printing where space is limited
- High resolution touch screen monitor with an intuitive menu structure makes the printer easy to learn and use
- Built-in tools for accuracy and workflow automation
- Fully integrated barcode scanner supports the ability to scan data from a request form or tissue sample barcode
- PathSmart Innovative Tracking Solution connects printers to a hub and delivers reporting data to help close gaps in chain of custody
- User login and tracking creates traceability for root cause analysis
- Intelligent slide selection prints the right type and number of slides based on the protocol selected for each tissue type
- Easy LIS integration and flexible connectivity - connects to any available on-market LIS
- Kit includes printer, 4 years and 1 year factory warranty of StatSwap service coverage, lifetime printhead guarantee, guaranteed LIS compatibility, guaranteed validated consumables (slides), PathSmart tracking system hub module, and 10 thermal transfer ribbons
Specifications
Specifications
Product Type | PiSmart S1 Printer |
For Use With (Equipment) | 10 Thermal Transfer Ribbons |