Learn More
StatLab PiSmart Slotted Laser Embedding Cassettes
Click to view available options
Color:
Blue
Gray
Green
Lavender
Orange
Pink
Tan
White
Yellow
Description
- 1,000 per box, 2 box per case, 2,000 per case
- Threaded in strips of 40 with lids separate
- PiSmart Cassettes contain 4 patented pips at all 4 corners on the front of each cassette, protecting the thermal printing from high heat during dewaxing
- Engineered specifically for use in the PiSmart Laser Cassette printer
- Available in 9 colors
Specifications
Specifications
Color | Lavender |
Product Type | Slotted Cassette |