Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIST Recombinant Rabbit Monoclonal Antibody (2B1P9)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA537819
Description
Positive Samples: HepG2, U-87MG, Rat brain, Mouse lung Immunogen sequence: MSAGGPCPAA AGGGPGGASC SVGAPGGVSM FRWLEVLEKE FDKAFVDVDL LLGEIDPDQA DITYEGRQKM TSLSSCFAQL CHKAQSVSQI NHKLEAQLVD.
PDZ domains contain approximately 90 amino acids and bind the extreme C terminus of proteins in a sequence-specific manner. PIST, a PDZ domain-containing Golgi protein, was discovered in a yeast two-hybrid system as a binding partner to Beclin-1, a Bcl-2-interacting protein homologous to the yeast autophagy gene apg6. Experiments with mutant PIST proteins lacking the PDZ domain showed that PIST interaction with Beclin-1 through its coiled-coil domain can modulate Beclin-1 activity and suggest that PIST interactions with other proteins through its PDZ domain may regulate the activity of PIST and Beclin-1.
Specifications
PIST | |
Recombinant Monoclonal | |
0.35 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BH60, Q9HD26 | |
GOPC | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-100 of human PIST/GOPC (Q9HD26). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
2B1P9 | |
Unconjugated | |
GOPC | |
2210402P09Rik; AI844555; CAL; CFTR-associated ligand; dJ94G16.2; dJ94G16.2 PIST; FIG; Fused in glioblastoma; golgi associated PDZ and coiled-coil motif containing; Golgi associated PDZ and coiled-coil motif containing protein; golgi-associated PDZ and coiled-coil motif containing protein; Golgi-associated PDZ and coiled-coil motif-containing protein; Gopc; GOPC1; PDZ domain-containing protein PIST; PDZ protein interacting specifically with TC10; PDZ/coiled-coil domain binding partner for the rho-family GTPase TC10; PIST | |
Rabbit | |
Affinity chromatography | |
RUO | |
309774, 57120, 94221 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction