Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ PIT-1 (POU1F1), Mouse, Unlabeled, Clone: 8B6.1,

Catalog No. MABN1602MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABN1602MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABN1602MI Supplier MilliporeSigma™ Supplier No. MABN1602
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pituitary-specific positive transcription factor 1 (UniProt P28069; also known as GHF-1, Growth hormone factor 1, PIT-1) is encoded by the POU1F1 (also known as CPHD1, GHF1, PIT1) gene (Gene ID 5449) in human. PIT-1 is a transcription factor that serves as a master regulator of lineage commitment and expansion during the anterior pituitary development process that drives regional expansion and functional differentiation of five distinct hormone-producing cell lineages, including somatotropes, lactotropes, thyrotropes, corticotropes, and gonadotropes. Loss of Pit-1 expression results in a combined pituitary hormone deficiency (CPHD) syndrome reflecting the absence of lactotropes, somatotropes, and a subset of thyrotropes. Although originally characterized as pituitary-specific, PIT-1 is also expressed in human breast. PIT-1 upregulation in breast tumors increases cell proliferation and the expression of two breast cancer-related hormones, growth hormone (GH) and prolactin (PRL), leading to MMPs upregulation and breast cancer metastasis. Human POU1F1 gene encodes two alternatively spliced isoforms, where Pit-1A (GHF-2) a.a. 47-73 is truncated to a single Thr47 residue in Pit-1B (GHF-1). Both PIT-1 isoforms contain a PIT-1 POU-specific domain (a.a. 150-224 of Pit-1A or a.a. 124-198 of Pit-1B) and a DNA-binding Homeobox (a.a. 240-299 of Pit-1A or a.a. 214-273 of Pit-1B).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIT-1 (POU1F1)
Applications Western Blot
Classification Monoclonal
Clone 8B6.1
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG2a in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Gene POU1F1, CPHD1, GHF1, PIT1
Gene Accession No. P28069
Host Species Mouse
Immunogen GST-tagged recombinant human PIT-1 internal fragment.
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_000297
Test Specificity Clone 8B6.1 epitope lies within an internal region present in both human PIT-1 spliced isoform A (Pit-1A, GHF-2) and isoform B (Pit-1B, GHF-1).
Target Species Human
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.