Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pit1 Antibody (PIT1/7262), Novus Biologicals™
SDP

Catalog No. p-200004714 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB234396 100 μg
NB235018 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB234396 Supplier Novus Biologicals Supplier No. NBP320299100UG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pit1 Monoclonal antibody specifically detects Pit1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen Pit1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PIT1/7262
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Immunohistochemistry-Paraffin 1:10-1:20
Formulation 10mM PBS with 0.05% BSA
Gene Alias CPHD1, GHF1, GHF-1pituitary-specific positive transcription factor 1, Growth hormone factor 1, PIT-1, PIT1pituitary-specific transcription factor 1, POU class 1 homeobox 1, POU domain class 1, transcription factor 1, POU domain, class 1, transcription factor 1, POU1F1a
Host Species Mouse
Immunogen Recombinant human full-length Pit1 protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Breast Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 5449
Target Species Human
Content And Storage Store at 4&drg;C. Do not freeze.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.