Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PIT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559662
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559662 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559662 Supplier Invitrogen™ Supplier No. PA559662
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CKLKAILSKW LEEAEQVGAL YNEKVGANER KRKRRTTISI AAKDALERHF GEQNKPSSQE IMRMAE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 97%.

PIT1 is a pituitary-specific transcription factor responsible for pituitary development and hormone expression in mammals and is a member of the POU family of transcription factors that regulate mammalian development. The POU family is so named because the first 3 members identified were PIT1 and OCT1 of mammals, and Unc-86 of C. elegans. PIT1 contains 2 protein domains, termed POU-specific and POU-homeo, which are both necessary for high affinity DNA binding on genes encoding growth hormone and prolactin. PIT1 is also important for regulation of the genes encoding prolactin and thyroid-stimulating hormone beta subunit by thyrotropin-releasing hormone and cyclic AMP.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PIT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene POU1F1
Gene Accession No. P28069
Gene Alias CPHD1; dw; dwarf; GHF1; GHF-1; GHF1A; growth hormone factor 1; Hmp1; homeodomain protein for Growth Hormone (GH); PIT1; pit-1; Pit1-rs1; PIT1Z; Pituary specific transcription factor 1 (growth hormone factor 1); pituitary specific transcription factor 1; pituitary specific transcription factor 1, related sequence 1; pituitary-specific positive transcription factor 1; pituitary-specific positive transcription factor 1 variant w; POU class 1 homeobox 1; POU domain, class 1, transcription factor 1; POU domain, class 1, transcription factor 1 (Pit1, growth hormone factor 1); POU1F1; POU1F1a; transcription factor Pit-1
Gene Symbols POU1F1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PIT1. Recombinant protein control fragment (Product #RP-98326).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5449
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.