Learn More
PIT2 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304474AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Sodium-phosphate symporter which seems to play a fundamental housekeeping role in phosphate transport by absorbing phosphate from interstitial fluid for normal cellular functions such as cellular metabolism, signal transduction, and nucleic acid and lipid synthesis. In vitro, sodium-dependent phosphate uptake is not siginificantly affected by acidic and alkaline conditions, however sodium-independent phosphate uptake occurs at acidic conditions. May play a role in extracellular matrix, cartilage and vascular calcification. Functions as a retroviral receptor and confers human cells susceptibility to infection to amphotropic murine leukemia virus (A-MuLV), 10A1 murine leukemia virus (10A1 MLV) and some feline leukemia virus subgroup B (FeLV-B) variants.Specifications
PIT2 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q08357, Q80UP8 | |
SLC20A2 | |
Between 300 and 350 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SLC20A2 | |
GLVR2, Glvr-2, MLVAR, PIT-2, Ram1, hPit2, pit2, gibbon ape leukemia virus receptor 2, murine leukemia virus, amphotropic, receptor for, phosphate transporter 2, sodium-dependent phosphate transporter 2, solute carrier family 20 member 2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20516, 6575 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.