Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PITHD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553519
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553519 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553519 Supplier Invitrogen™ Supplier No. PA553519
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: RSKFVESDAD EELLFNIPFT GNVKLKGIII MGEDDDSHPS EMRLYKNIPQ MSFDDTEREP DQTFSLNRDL TGELEYATKI SRFSNVYHLS IHISKNFGAD TTKVFYIGLR GEWTELRRHE VTICNYEASA NPA Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 76%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PITHD1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PITHD1
Gene Accession No. Q9GZP4
Gene Alias AD039; C1orf128; HT014; PITH (C-terminal proteasome-interacting domain of thioredoxin-like) domain containing 1; PITH domain containing 1; PITH domain-containing protein 1; PITHD1; PP603; TXNL1 C-terminal like; TXNL1CL
Gene Symbols PITHD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PITHD1. Recombinant protein control fragment (Product #RP-89829).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57095
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.