Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PITPN Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104010
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104010 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104010 Supplier Invitrogen™ Supplier No. PA5104010
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PITPNA.

This gene encodes a member of a family of lipid-binding proteins that transfer molecules of phosphatidylinositol or phosphatidylcholine between membrane surfaces. The protein is implicated in phospholipase C signaling and in the production of phosphatidylinositol 3,4,5-trisphosphate (PIP3) by phosphoinositide-3-kinase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PITPN
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PITPNA
Gene Accession No. P16446, P53810, Q00169
Gene Alias epididymis secretory protein Li 36; HEL-S-36; phosphatidylinositol transfer protein alpha; phosphatidylinositol transfer protein alpha isoform; phosphatidylinositol transfer protein, alpha; Pitp alpha; PI-TPalpha; PI-TP-alpha; PITPN; Pitpna; PITPNB; ptdIns transfer protein alpha; ptdInsTP alpha; vb; VIB1A; vibrator
Gene Symbols PITPNA
Host Species Rabbit
Immunogen A synthesized peptide derived from human PITPNA(Accession Q00169), corresponding to amino acid residues L150-F200.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18738, 29525, 5306
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.