Learn More
Invitrogen™ PITPNB Monoclonal Antibody (OTI6H10)

Description
PITPNB Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | PITPNB |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI6H10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PITPNB |
| Gene Accession No. | P48739 |
| Gene Alias | phosphatidylinositol transfer protein beta; phosphatidylinositol transfer protein beta isoform; phosphatidylinositol transfer protein, beta; phosphotidylinositol transfer protein, beta; Pitp beta; PI-TP beta; PI-TP-beta; Pitpnb; ptdIns transfer protein beta; PtdInsTP; ptdInsTP beta; VIB1B |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.