Learn More
PITPNB Mouse anti-Human, Clone: OTI6H10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF809305

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The lipid binding proteins known as phosphatidylinositol transfer proteins (PITP) facilitate the formation of phosphatidylinositol derived second messenger molecules, which are related to the phospholipase C and phosphoinositide 3-kinase pathways. PITP are ubiquitously expressed proteins that transfer phosphatidylinositol (PI) and phosphatidylcholine (PC) between membranes enriched in PI or PC to membranes that are deficient in PI or PC. PITP mobilizes PI from the endoplasmic recticulum and regulates the release of PI from stored vesicles in the Golgi network. In mammalian cells, three smaller forms of soluble PITP are present, designated PITP alpha, PITP beta and retinal degeneration B (rdgB) beta. PITP beta is a 271 amino acid protein that is widely expressed in various tissues. Though required for Golgi targeting, constitutive phosphorylation of Ser-262 has no effect on phospholipid transfer activity. There are two isoforms of PITP beta that are produced as a result of alternative splicing events.Specifications
PITPNB | |
Monoclonal | |
Unconjugated | |
PITPNB | |
PI-TP-beta, PtdInsTP, VIB1B | |
Mouse | |
Affinity Chromatography | |
RUO | |
23760 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunohistochemistry (Paraffin), Western Blot | |
OTI6H10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P48739 | |
PITPNB | |
Full length human recombit protein of human PITPNB produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.