Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PITPNB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA551403
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA551403 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA551403 Supplier Invitrogen™ Supplier No. PA551403
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CSVQEYQVGQ LYSVAEASKN ETGGGEGIEV LKNEPYEKDG EKGQYTHKIY HLKSKVPAFV RMIAPEGSLV FHEKAWNAYP YCRTIVTNEY MKDDFFIKIE TWHKPDLGTL ENVHGLDPNT Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.

This gene encodes a cytoplasmic protein that catalyzes the transfer of phosphatidylinositol and phosphatidylcholine between membranes. This transfer activity is required for COPI complex-mediated retrograde transport from the Golgi apparatus to the endoplasmic reticulum. Alternative splicing of this gene results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PITPNB
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PITPNB
Gene Accession No. P48739, P53811, P53812
Gene Alias phosphatidylinositol transfer protein beta; phosphatidylinositol transfer protein beta isoform; phosphatidylinositol transfer protein, beta; phosphotidylinositol transfer protein, beta; Pitp beta; PI-TP beta; PI-TP-beta; Pitpnb; ptdIns transfer protein beta; PtdInsTP; ptdInsTP beta; VIB1B
Gene Symbols PITPNB
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PITPNB. Recombinant protein control fragment (Product #RP-91391).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 114561, 23760, 56305
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.