Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PITPNC1 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297834AF700
Description
PITPNC1 Polyclonal specifically detects PITPNC1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
PITPNC1 | |
Polyclonal | |
50 mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
26207 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 700 | |
cytoplasmic phosphatidylinositol transfer protein 1, Mammalian rdgB homolog beta, M-rdgB beta, MrdgBbeta, phosphatidylinositol transfer protein, cytoplasmic 1, RDGBB, RDGBB1, rdgBbeta, RDGB-BETA, retinal degeneration B beta 1, Retinal degeneration B homolog beta | |
Produced in rabbits immunized with E. coli-derived Human PITPNC1 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction