Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PITPNC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA530508
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA530508 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA530508 Supplier Invitrogen™ Supplier No. PA530508
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HepG2. Predicted reactivity: Mouse (97%), Pig (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the phosphatidylinositol transfer protein family. The encoded cytoplasmic protein transfers phosphatidylinositol from one membrane compartment to another. Two transcript variants encoding distinct isoforms have been identified for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PITPNC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene PITPNC1
Gene Accession No. Q9UKF7
Gene Alias 1110020B03Rik; 5830436L09Rik; AI662802; AI851387; C330017I21Rik; cytoplasmic phosphatidylinositol transfer protein 1; diabetic nephropathy-related protein; Dnr411; Mammalian rdgB homolog beta; mM-rdgBbeta; M-rdgB beta; mrdgBbeta; M-RDGB-beta; phosphatidylinositol transfer protein; phosphatidylinositol transfer protein cytoplasmic 1; phosphatidylinositol transfer protein cytoplasmic 1a; phosphatidylinositol transfer protein, cytoplasmic 1; pitpnc1; pitpnc1a; RDGBB; RDGBB1; RdgBbeta; RDGB-BETA; retinal degeneration B beta; retinal degeneration B beta 1; retinal degeneration B homolog beta
Gene Symbols PITPNC1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PITPNC1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26207
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.