Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PITPNM3 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NB300814
Description
PITPNM3 Polyclonal specifically detects PITPNM3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.Specifications
PITPNM3 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
membrane-associated phosphatidylinositol transfer protein 3, MGC157740, MGC157741, NIR-1, NIR1CORD5, Phosphatidylinositol transfer protein, membrane-associated 3, PITPnm 3, PITPNM family member 3, Pyk2 N-terminal domain-interacting receptor 1, RDGBA3, retinal degeneration B alpha 3 | |
Goat | |
Affinity Purified | |
RUO | |
83394 | |
Human | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA | |
0.5 mg/mL | |
Immunohistochemistry 5 - 10 ug/ml, Immunohistochemistry-Paraffin 5 - 10 ug/ml, Peptide ELISA Detection limit 1:16000 | |
Q9BZ71 | |
PITPNM3 | |
Peptide with sequence C-SWARGPPKFESVP corresponding to C-Terminus according to NP_112497.2, NP_001159438.1. | |
0.1 mg | |
Primary | |
Human. Predicted cross-reactivity based on sequence identity: Mouse, Dog. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction