Learn More
Invitrogen™ PITPNM3 Polyclonal Antibody

Description
Immunogen sequence: RFPLGDGQSL LLADALHTHS PLFLEGSSRD SPPLLDAPAS PPQASRFQRP GRRMSEGSSH SESSESSDSM APVGASRITA KWWGSKRIDY ALYCPDV Highest antigen sequence identity to the following orthologs: Mouse - 90%, Rat - 91%.
Specifications
Specifications
| Antigen | PITPNM3 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | PITPNM3 |
| Gene Accession No. | Q9BZ71 |
| Gene Alias | A330068P14Rik; ACKR6; AI848332; atypical chemokine receptor 6; cone rod dystrophy 5; CORD5; Gm880; Membrane-associated phosphatidylinositol transfer protein 3; MGC157740; MGC157741; NIR1; NIR-1; phosphatidylinositol transfer protein, membrane-associated 3; PITPnm 3; PITPNM family member 3; Pitpnm3; Pyk2 N-terminal domain-interacting receptor 1; RDGBA3; retinal degeneration B alpha 3 |
| Gene Symbols | PITPNM3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.