Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PITRM1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552339
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552339 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552339 Supplier Invitrogen™ Supplier No. PA552339
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELRSQQSKPQ DASCLPALKV SDIEPTIPVT ELDVVLTAGD IPVQYCAQPT NGMVYFRAFS SLNTLPEELR PYVPLFCSVL TKLGCGLLDY REQAQQIELK TGGMSASPHV LPDDSHMDTY EQGVLFSSLC LDRNLPDMMQ LWSEI Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 83%.

The protein encoded by this gene is an ATP-dependent metalloprotease that degrades post-cleavage mitochondrial transit peptides. The encoded protein binds zinc and can also degrade amyloid beta A4 protein, suggesting a possible role in Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PITRM1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PITRM1
Gene Accession No. Q5JRX3
Gene Alias 2310012C15Rik; AA410010; hPreP; KIAA1104; Metalloprotease 1; metalloprotease 1 (pitrilysin family); mKIAA1104; MP1; MP-1; Ntup1; nuclear transplantation upregulated protein 1; pitrilysin metallepetidase 1; pitrilysin metallopeptidase 1; pitrilysin metalloprotease 1; pitrilysin metalloproteinase 1; Pitrm1; PREP; PreP peptidasome; presequence protease, mitochondrial
Gene Symbols PITRM1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PITRM1. Recombinant protein control fragment (Product #RP-89248).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10531
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.