Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PITX1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP188644
Description
PITX1 Polyclonal specifically detects PITX1 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.Specifications
PITX1 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Frozen Reported in scientific literature (PMID: 24909994). | |
P78337 | |
PITX1 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:SFTFFNSMSPLSSQSMFSAPSSISSMTMPSSMGPGAVPGMPNSGLNNINNLTGSSLNSAMSPGACPYGTPASPYSVYRDTCNSSLASLRLKSKQHSS | |
0.1 mL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
Unconjugated | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
BFTpituitary homeobox 1, CCF, Hindlimb-expressed homeobox protein backfoot, Homeobox protein PITX1, paired-like homeodomain 1, pituitary otx-related factor, POTXpituitary homeo box 1, PTX1Paired-like homeodomain transcription factor 1hindlimb expressed homeobox protein backfoot | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
5307 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction