Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PITX1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101136
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101136 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101136 Supplier Thermo Scientific Supplier No. PA5101136
Add to Cart
Add to Cart

PITX1 Polyclonal Antibody

PITX1 Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen PITX1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PITX1
Gene Accession No. P70314, P78337, Q99NA7
Gene Alias BFT; CCF; hindlimb expressed homeobox protein backfoot; hindlimb-expressed homeobox protein backfoot; homeobox protein PITX1; Homeobox protein P-OTX; LBNBG; OTX; paired like homeodomain 1; paired-like homeodomain 1; paired-like homeodomain transcription factor 1; pituitary homeo box 1; pituitary homeobox 1; Pituitary OTX-related factor; Pitx1; Potx; P-OTX; PTX1; Unknown (protein for MGC:136976); zgc:136976
Gene Symbols PITX1
Host Species Rabbit
Immunogen A synthesized peptide derived from human PITX1(Accession P78337), corresponding to amino acid residues R110-Y160.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113983, 18740, 5307
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.