Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PITX2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA565596
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA565596 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA565596 Supplier Invitrogen™ Supplier No. PA565596
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ACVQLGVQPAA VECLFSKDSE IKKVEFTDSP ESRKEAASSK FFPRQHPGAN EKDKSQQGKN EDVGAEDPSK KKR Highest antigen sequence identity to the following orthologs - mouse 95%, rat 31%.

Pilx2 is a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. This protein acts as a transcription factor and regulates procollagen lysyl hydroxylase gene expression. It plays a role in the terminal differentiation of somatotroph and lactotroph cell phenotypes, is involved in the development of the eye, tooth and abdominal organs, and acts as a transcriptional regulator involved in basal and hormone-regulated activity of prolactin. Mutations in this protein are associated with Axenfeld-Rieger syndrome, iridogoniodysgenesis syndrome, and sporadic cases of Peters anomaly. A similar protein in other vertebrates is involved in the determination of left-right asymmetry during development.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PITX2
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PITX2
Gene Accession No. Q99697
Gene Alias 9430085M16Rik; all1-responsive gene 1; ALL1-responsive protein ARP1; Arp1; Brx1; BRX1 homeoprotein; Brx1b; Homeobox protein PITX2; homeodomain transcription factor 2; IDG2; IGDS; IGDS2; IHG2; IRID2; Munc30; orthodenticle-like homeobox 2; Otlx2; paired like homeodomain 2; paired like homeodomain transcription factor 2; paired-like homeodomain 1; paired-like homeodomain 2; paired-like homeodomain transcription factor 1; paired-like homeodomain transcription factor 2; paired-like homeodomain transcription factor 2c; paired-like homeodomain transcription factor Munc 30; Pituitary homeobox 2; PITX1; Pitx2; Pitx-2; Pitx2c; Ptx2; Rgs; Rieg; RIEG bicoid-related homeobox transcription factor; rieg bicoid-related homeobox transcription factor 1; RIEG1; rPtx2; RS; S a 1 glycoprotein; S a1glycoprotein; S alpha 1 glycoprotein; S α 1 glycoprotein; S α1glycoprotein; Solurshin
Gene Symbols PITX2
Host Species Rabbit
Immunogen Recombinant Human PITX2. Recombinant protein control fragment (Product #RP-106258).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5308
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.