Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PITX3 Antibody (28A9), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32638050UL
Description
PITX3 Monoclonal antibody specifically detects PITX3 in Human samples. It is validated for Flow Cytometry, ELISA, ImmunocytochemistrySpecifications
PITX3 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, ELISA, Immunocytochemistry | |
28A9 | |
Flow Cytometry 1:50-1:200, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
CTPP4, Homeobox protein PITX3, MGC12766, paired-like homeodomain 3, Paired-like homeodomain transcription factor 3, pituitary homeobox 3, PTX3 | |
A synthesized peptide derived from Human PITX3 [UniProt O75364] | |
50 μL | |
Neurodegeneration, Neuroscience | |
5309 | |
Store at -20 to -70C. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction