Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PITX3 Antibody (SR1536), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | PITX3 |
---|---|
Clone | SR1536 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunoprecipitation 1:50 |
Applications | Western Blot, Flow Cytometry, Immunoprecipitation |
Classification | Monoclonal |
Description
PITX3 Monoclonal antibody specifically detects PITX3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, ImmunoprecipitationSpecifications
PITX3 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CTPP4, Homeobox protein PITX3, MGC12766, paired-like homeodomain 3, Paired-like homeodomain transcription factor 3, pituitary homeobox 3, PTX3 | |
A synthesized peptide derived from human PITX3 (Uniprot #: O75364) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1536 | |
Western Blot, Flow Cytometry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Neurodegeneration, Neuroscience | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
5309 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title