Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PITX3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587989
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587989 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587989 Supplier Invitrogen™ Supplier No. PA587989
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MEFGLLSEAE ARSPALSLSD AGTPHPQLPE HGCKGQEHSD SEKASASLPG GSPEDGSLKK KQRRQRTHFT; Positive Samples: 293T, U-87MG, Mouse heart, Rat kidney, Rat brain; Cellular Location: Nucleus.

This gene encodes a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. Members of this family act as transcription factors. This protein is involved in lens formation during eye development. Mutations of this gene have been associated with anterior segment mesenchymal dysgenesis and congenital cataracts.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PITX3
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.72 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PITX3
Gene Accession No. O35160, O75364, P81062
Gene Alias ak; aphakia; ASMD; ASOD; CTPP4; CTRCT11; Homeobox protein PITX3; homeobox protein PTX3; MGC12766; paired like homeodomain 3; paired-like homeodomain 3; paired-like homeodomain transcription factor 3; Pituitary homeobox 3; Pituitary homeobox 3 (Homeobox protein PITX3); PITX3; Ptx3
Gene Symbols PITX3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-70 of human PITX3 (NP_0050201).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18742, 29609, 5309
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.