Learn More
Invitrogen™ PITX3 Recombinant Rabbit Monoclonal Antibody (3P4U8)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA542696
Description
Positive test controls include: C6, SH-SY5Y, U-87MG, Mouse eye. The target is usually found in the following locations: Nucleus. Immunogen sequence: MEFGLLSEAE ARSPALSLSD AGTPHPQLPE HGCKGQEHSD SEKASASLPG GSPEDGSLKK KQRRQRTHFT SQQLQELEAT FQRNRYPDMS TREEIAVWTN.
This gene encodes a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. Members of this family act as transcription factors. This protein is involved in lens formation during eye development. Mutations of this gene have been associated with anterior segment mesenchymal dysgenesis and congenital cataracts.
Specifications
PITX3 | |
Recombinant Monoclonal | |
0.5 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35160, O75364, P81062 | |
PITX3 | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human PITX3 (O75364). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
ELISA, Western Blot | |
3P4U8 | |
Unconjugated | |
PITX3 | |
ak; aphakia; ASMD; ASOD; CTPP4; CTRCT11; Homeobox protein PITX3; homeobox protein PTX3; MGC12766; paired like homeodomain 3; paired-like homeodomain 3; paired-like homeodomain transcription factor 3; Pituitary homeobox 3; Pituitary homeobox 3 (Homeobox protein PITX3); PITX3; Ptx3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
18742, 29609, 5309 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.