Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pivaldehyde, 96%

Catalog No. p-2692067
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 630-19-3 | C5H10O | 86.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 630-19-3
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00006962
InChI Key FJJYHTVHBVXEEQ-UHFFFAOYSA-N
Synonym trimethylacetaldehyde, pivalaldehyde, pivaldehyde, propanal, 2,2-dimethyl, neopentanal, pivalic aldehyde, 2,2-dimethylpropionaldehyde, unii-ssc20260qw, trimethyl acetaldehyde, trimethylacetaldehyde pivaldehyde
PubChem CID 12417
IUPAC Name 2,2-dimethylpropanal
SMILES CC(C)(C)C=O

Specifications

Melting Point 6.0°C
Color Colorless
Density 0.7750g/mL
Boiling Point 75.0°C
Flash Point −15°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.3775 to 1.3795
Linear Formula (CH3)3CCHO
Quantity 5 g
Beilstein 01, IV, 3295
Fieser 13,249
Specific Gravity 0.775
Merck Index 15, 7624
Solubility Information Solubility in water: negligibly soluble.
Formula Weight 86.13
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material Pivaldehyde, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Highly flammable liquid and vapor.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-134-6

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.