Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 3-trifluoromethylbenzoylacetate, 95%, Thermo Scientific™

Catalog No. AC425450025
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle

Chemical Identifiers

CAS 93618-66-7
Molecular Formula C11H9F3O3
Molecular Weight (g/mol) 246.19
MDL Number MFCD00216522
InChI Key RPRMYRPHNDGZOY-UHFFFAOYSA-N
Synonym methyl 3-oxo-3-3-trifluoromethyl phenyl propanoate, methyl 3-trifluoromethyl benzoylacetate, methyl 3-trifluoromethyl benzoyl acetate, 3-3-trifluoromethylphenyl-3-oxo-propionic acid methyl ester, 3-oxo-3-3-trifluoromethyl-phenyl-propionic acid methyl ester, pubchem2695, methyl 3-trifluoromethylbenzoylacetate, methyl 3-oxo-3-3-trifluoromethylphenyl propanoate, 3-oxo-3-3-trifluoromethylphenyl propionic acid methyl ester
PubChem CID 735880
IUPAC Name methyl 3-oxo-3-[3-(trifluoromethyl)phenyl]propanoate
SMILES COC(=O)CC(=O)C1=CC=CC(=C1)C(F)(F)F

Specifications

Color Yellow
Boiling Point 94°C to 96°C (0.2 mmHg)
Flash Point >200°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Refractive Index 1.4864 to 1.4884
Quantity 2.5 g
Solubility Information Solubility in water: neglegible in water
Formula Weight 246.18
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material Methyl 3-trifluoromethylbenzoylacetate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.