Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Piwil/Mili Polyclonal Antibody, Biotin, Bioss
SDP

Catalog No. 502281441
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-228-1441 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-228-1441 Supplier Bioss Supplier No. BS3817RBIOTIN
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Piwil/Mili Polyclonal antibody specifically detects Piwil/Mili in Human, Mouse, Rat samples. It is validated for Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Piwil/Mili
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Biotin
Formulation TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300
Gene Piwil2
Gene Accession No. Q8CDG1, Q8TC59
Gene Alias 80kDa PIWIL2 short isoform; cancer/testis antigen 80; CT80; HILI; mili; Miwi like; Miwi likw; piwi like homolog 2; piwi like RNA-mediated gene silencing 2; PIWIL1L; Piwil2; PIWI-L2; piwil2-like protein; piwi-like 2; piwi-like homolog 2; piwi-like protein 2; piwi-like RNA-mediated gene silencing 2; testicular tissue protein Li 141
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human Piwil/Mili
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 306011, 55124, 57746
Target Species Human, Mouse, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.