Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIWIL1/HIWI Rabbit anti-Mouse, Rat, Clone: 0H5A4, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP31620020UL
This item is not returnable.
View return policy
Description
PIWIL1/HIWI Monoclonal antibody specifically detects PIWIL1/HIWI in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
PIWIL1/HIWI | |
Monoclonal | |
Unconjugated | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
Rabbit | |
Affinity purified | |
RUO | |
9271 | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
0H5A4 | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin | |
HIWIpiwi homolog, MIWI, PIWI, piwi (Drosophila)-like 1, piwi-like 1 (Drosophila), piwi-like protein 1 | |
A synthetic peptide corresponding to a sequence within amino acids 700-800 of human PIWIL1/HIWI (Q96J94). | |
20 μg | |
Primary | |
Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction