Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PIWIL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA517034
Description
It is not recommended to aliquot this antibody.
Hiwi is a member of the Piwi /Argonaute protein family. Hiwi is specifically expressed in germline cells such as spermatocytes and round spermatids during spermatogenesis. Hiwi is also expressed in CD34+ hematopoietic progenitors, but not in CD34- hematopoietic cells, suggesting a role for this gene in the maintenance of hematopoietic stem cells.
Specifications
PIWIL1 | |
Polyclonal | |
Unconjugated | |
PIWIL1 | |
CT80; CT80.1; HILI; HIWI; mili; Miwi; Miwi like; Miwi likw; PIWI; piwi homolog; piwi like homolog 1; piwi like RNA-mediated gene silencing 1; PIWIL1; PIWIL1L; piwil2-like protein; piwi-like 1; piwi-like homolog 1; piwi-like protein 1; piwi-like protein 2; piwi-like RNA-mediated gene silencing 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
363912, 57749, 9271 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot, Immunocytochemistry, Western Blot | |
85 μg/mL | |
0.01M HEPES with 100 μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5 | |
Q96J94, Q9JMB7 | |
PIWIL1 | |
Synthetic peptide corresponding to the sequence surrounding Gly82 of mouse Miwi. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction